Gene Transcripts

Gene Symbol
Wdr45
Entrez Gene ID
54636
Gene Name
WD repeat domain 45
Gene Aliases
C79260, DXImx38e, JM5, Sfc19, WIPI-4, Wdrx1
Chromosome Location
Chr. X: 7721995 - 7728201 on Build GRCm38
UniGene ID
Mm.305725
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290792.1 NP_001277721.1 6 543
NM_001290794.1 NP_001277723.1 6 439
NM_001290795.1 NP_001277724.1 6 543
NM_172372.2 NP_758960.1 6 539
GenBank mRNA AF229635.1 - 6 517
AK018409.1 - 6 539
AK145616.1 - 6 439
BC011479.1 - 6 510 4035468

View Full Product Details