Gene Transcripts

Gene Symbol
Wnt7a
Entrez Gene ID
22421
Gene Name
wingless-type MMTV integration site family, member 7A
Gene Aliases
AI849442, Wnt-7a, px, tw
Chromosome Location
Chr. 6: 91363983 - 91411369 on Build GRCm38
UniGene ID
Mm.56964
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009527.3 NP_033553.2 4 1852
GenBank mRNA AK004683.1 - 4 1853
BC049093.1 - 4 1846 6415801
BC057586.1 - 4 1846 6415801

View Full Product Details