Gene Transcripts

Gene Symbol
Trip6
Entrez Gene ID
22051
Gene Name
thyroid hormone receptor interactor 6
Gene Aliases
Chromosome Location
Chr. 5: 137309899 - 137314487 on Build GRCm38
UniGene ID
Mm.27063
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011639.3 NP_035769.1 9 1611
GenBank mRNA AF097511.1 - 9 1504
AF116823.1 - 9 1606
AK010435.1 - 9 1612
AK146823.1 - 9 1561
AK159387.1 - 9 1580
AK166024.1 - 9 1776
AK204151.1 - 1 78
BC138685.1 - 9 1543 8861706
BC138686.1 - 9 1543 8861707

View Full Product Details