Gene Transcripts

Gene Symbol
Katna1
Entrez Gene ID
23924
Gene Name
katanin p60 (ATPase-containing) subunit A1
Gene Aliases
Chromosome Location
Chr. 10: 7726000 - 7763150 on Build GRCm38
UniGene ID
Mm.28127
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011835.2 NP_035965.2 3 332
GenBank mRNA AF153197.1 - 3 310
AK012319.1 - 3 332
AK041900.1 - 3 327
BC009136.1 - 3 246 3597742

View Full Product Details