Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001291033.1 | NP_001277962.1 | 4 | 1111 | |
NM_001291034.1 | NP_001277963.1 | 3 | 1186 | ||
NM_001291035.1 | NP_001277964.1 | 3 | 1327 | ||
NM_001291036.1 | NP_001277965.1 | 4 | 1176 | ||
NM_001291037.1 | NP_001277966.1 | 3 | 1425 | ||
NM_011841.2 | NP_035971.1 | 4 | 1061 | ||
GenBank mRNA | AB019373.1 | - | 4 | 1011 | |
AF126159.1 | - | 3 | 824 | ||
AF126160.1 | - | 3 | 874 | ||
AF126161.1 | - | 3 | 1244 | ||
AK148119.1 | - | 4 | 1043 | ||
AK155187.1 | - | 4 | 979 | ||
AY534740.1 | - | 4 | 995 | ||
BC013745.1 | - | 3 | 1344 | ||
BC033598.1 | - | 4 | 1151 | ||
BC051020.1 | - | 2 | 1705 | ||
BC058096.1 | - | 2 | 1705 | ||
BC082315.1 | - | 3 | 1248 | ||
BC100398.1 | - | 3 | 1128 | 30470655 |