Gene Transcripts

Gene Symbol
Tenm4
Entrez Gene ID
23966
Gene Name
teneurin transmembrane protein 4
Gene Aliases
Doc4, ELM2, Odz4, R75022, Ten-m4, l(7)-3Rn, l7Rn3, mKIAA1302
Chromosome Location
Chr. 7: 96171237 - 96911093 on Build GRCm38
UniGene ID
Mm.254610
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011858.3 NP_035988.2 19 3206
GenBank mRNA AB025413.1 - 17 3039
AF059485.1 - 20 3617
AK039472.1 - 5 738
AK147329.1 - 19 3207
AK147579.1 - 19 3506

View Full Product Details