Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_012053.2 | NP_036183.1 | 2 | 171 | |
GenBank mRNA | AK088263.1 | - | 2 | 158 | |
AK131970.1 | - | 2 | 163 | ||
AK145498.1 | - | 2 | 158 | ||
AK146330.1 | - | 2 | 158 | ||
AK151408.1 | - | 2 | 158 | ||
AK152063.1 | - | 2 | 165 | ||
AK167775.1 | - | 2 | 158 | ||
AK168332.1 | - | 2 | 158 | ||
AK168769.1 | - | 2 | 156 | ||
BC043017.1 | - | 1 | 139 | 5684024 | |
U67771.1 | - | 1 | 143 |