Gene Transcripts

Gene Symbol
Nrk
Entrez Gene ID
27206
Gene Name
Nik related kinase
Gene Aliases
AA407323, AA409510, AI225895, AV168563, AW538185, Nesk
Chromosome Location
Chr. X: 138914312 - 139010532 on Build GRCm38
UniGene ID
Mm.482319
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013724.2 NP_038752.2 8 996
GenBank mRNA AB020741.1 - 8 921
AB035267.1 - 8 674
AK041377.1 - 8 996
BC068311.1 - 8 971
BC139029.1 - 8 763 8862051
BC139031.1 - 8 763 8862053

View Full Product Details