Gene Transcripts

Gene Symbol
Tpst1
Entrez Gene ID
22021
Gene Name
protein-tyrosine sulfotransferase 1
Gene Aliases
R75054, Tango13a
Chromosome Location
Chr. 5: 130073326 - 130135734 on Build GRCm38
UniGene ID
Mm.332720
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130476.2 NP_001123948.1 6 1913
NM_013837.2 NP_038865.1 6 1730
GenBank mRNA AF038008.1 - 6 1730
AK150753.1 - 6 1798
BB805916.1 - 1 237
BC012666.1 - 6 1660 4035106

View Full Product Details