Gene Transcripts
- Gene Symbol
- Clec5a
- Entrez Gene ID
- 23845
- Gene Name
- C-type lectin domain family 5, member a
- Gene Aliases
- Clecsf5, Ly100, MDL-1, Mdl1
- Chromosome Location
- Chr. 6: 40574898 - 40585805 on Build GRCm38
- UniGene ID
- Mm.103765
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001038604.1 |
NP_001033693.1
|
4, 5 |
390 |
|
NM_021364.2 |
NP_067339.1
|
3, 4 |
315 |
|
GenBank mRNA |
AF139769.1 |
- |
3, 4 |
285 |
|
AK036697.1 |
- |
3, 4 |
281 |
|
AK046600.1 |
- |
3, 4 |
302 |
|
AK137482.1 |
- |
3, 4 |
316 |
|
BC104364.1 |
- |
4, 5 |
349 |
40049817 |
BC104365.1 |
- |
4, 5 |
349 |
40049818 |
BY205833.1 |
- |
3, 4 |
331 |
|
CN670295.1 |
- |
4, 5 |
352 |
|
View Full Product Details