Gene Transcripts

Gene Symbol
Bves
Entrez Gene ID
23828
Gene Name
blood vessel epicardial substance
Gene Aliases
Pop1, Popdc1, mBVES
Chromosome Location
Chr. 10: 45335762 - 45372484 on Build GRCm38
UniGene ID
Mm.332793
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024285.2 NP_077247.1 5, 6 773
GenBank mRNA AF124510.1 - 4, 5 634
AF204174.1 - 5, 6 771
BC132018.1 - 4, 5 663 40130295
BC132044.1 - 4, 5 663 40130321

View Full Product Details