Gene Transcripts

Gene Symbol
Nr5a1
Entrez Gene ID
26423
Gene Name
nuclear receptor subfamily 5, group A, member 1
Gene Aliases
Ad4BP, ELP, ELP-3, Ftz-F1, Ftzf1, SF-1, SF1
Chromosome Location
Chr. 2: 38692656 - 38714560 on Build GRCm38
UniGene ID
Mm.31387
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_139051.3 NP_620639.1 7 1532
GenBank mRNA AB000490.1 - 7 1579
AF511594.1 - 7 1362
AK134378.1 - 7 1533
AK144376.1 - 7 1493
AK156911.1 - 7 1500
BC110476.1 - 7 1475 40047261
BC110477.1 - 7 1475 40047262

View Full Product Details