Gene Transcripts

Gene Symbol
Thbd
Entrez Gene ID
83580
Gene Name
thrombomodulin
Gene Aliases
Chromosome Location
Chr. 3: 142748673 - 142752325 on Build Rnor_6.0
UniGene ID
Rn.88295
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031771.2 NP_113959.1 1 388
GenBank mRNA AF022743.1 - 1 313
BC070903.1 - 1 388 7100705

View Full Product Details