Gene Transcripts
- Gene Symbol
- ZPBP
- Entrez Gene ID
- 11055
- Gene Name
- zona pellucida binding protein
- Gene Aliases
- ZPBP1
- Chromosome Location
- Chr. 7: 49846355 - 50093264 on Build GRCh38
- UniGene ID
- Hs.388841
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001159878.1 |
NP_001153350.1
|
6, 7 |
858 |
|
NM_007009.2 |
NP_008940.2
|
6, 7 |
861 |
|
XM_011515095.2 |
XP_011513397.1
|
6, 7 |
855 |
|
XM_011515096.2 |
XP_011513398.1
|
6, 7 |
830 |
|
XM_011515097.1 |
XP_011513399.1
|
6, 7 |
855 |
|
XM_011515100.2 |
XP_011513402.1
|
5, 6 |
683 |
|
XM_011515101.2 |
XP_011513403.1
|
5, 6 |
656 |
|
XM_011515102.2 |
XP_011513404.1
|
6, 7 |
1034 |
|
XM_017011707.1 |
XP_016867196.1
|
6, 7 |
855 |
|
GenBank mRNA |
AK312799.1 |
- |
6, 7 |
830 |
|
BC005223.1 |
- |
6, 7 |
820 |
3950015 |
BT006822.1 |
- |
6, 7 |
791 |
|
D17570.1 |
- |
6, 7 |
827 |
|
View Full Product Details