Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_007083.4 | NP_009014.2 | 2 | 307 | |
NM_198041.2 | NP_932158.1 | 2 | 239 | ||
GenBank mRNA | AB209758.1 | - | 1 | 4926 | |
AF019632.2 | - | 2 | 203 | ||
AF019633.2 | - | 2 | 307 | ||
AK291871.1 | - | 2 | 306 | ||
AK294450.1 | - | 2 | 239 | ||
BC009842.2 | - | 2 | 287 | 3942416 | |
BG386740.1 | - | 2 | 285 | ||
CB128685.1 | - | 2 | 308 |