Gene Transcripts
- Gene Symbol
- Pnliprp2
- Entrez Gene ID
- 117554
- Gene Name
- pancreatic lipase-related protein 2
- Gene Aliases
- Chromosome Location
- Chr. 1: 279896973 - 279927600 on Build Rnor_6.0
- UniGene ID
- Rn.87688
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_057206.1 |
NP_476554.1
|
3 |
355 |
|
GenBank mRNA |
L09216.1 |
- |
3 |
355 |
|
View Full Product Details