Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001300791.1 | NP_001287720.1 | 3 | 480 | |
NM_001300792.1 | NP_001287721.1 | 3 | 480 | ||
NM_007054.6 | NP_008985.3 | 3 | 480 | ||
XM_006714526.3 | XP_006714589.1 | 3 | 535 | ||
XM_017008995.1 | XP_016864484.1 | 3 | 535 | ||
XM_017008996.1 | XP_016864485.1 | 3 | 535 | ||
GenBank mRNA | AB209780.1 | - | 3 | 428 | |
AF041853.1 | - | 2 | 321 | ||
AK295089.1 | - | 3 | 438 | ||
AK313359.1 | - | 3 | 448 | ||
AM177178.1 | - | 2 | 321 | ||
BC020890.1 | - | 3 | 445 | ||
BC032599.1 | - | 3 | 463 | ||
BC039592.1 | - | 3 | 437 | ||
BC045542.1 | - | 3 | 474 | 5298675 | |
BI561509.1 | - | 3 | 480 |