Gene Transcripts

Gene Symbol
Pga5
Entrez Gene ID
60372
Gene Name
pepsinogen 5, group I
Gene Aliases
Pepf
Chromosome Location
Chr. 1: 226722198 - 226732736 on Build Rnor_6.0
UniGene ID
Rn.43875
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021753.1 NP_068521.1 2 129
GenBank mRNA AJ251687.1 - 2 129

Gene Symbol
LOC100910889
Entrez Gene ID
100910889
Gene Name
pepsin F-like
Gene Aliases
Chromosome Location
Chr. Un|NW_007906915: 96133 - 107472 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003749058.3 XP_003749106.1 2 788

View Full Product Details