Gene Transcripts

Gene Symbol
Chrna10
Entrez Gene ID
64574
Gene Name
cholinergic receptor, nicotinic, alpha 10 (neuronal)
Gene Aliases
Chromosome Location
Chr. 1: 167206722 - 167210919 on Build Rnor_6.0
UniGene ID
Rn.48767
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022639.1 NP_072161.1 5 1659
XM_006229891.2 XP_006229953.1 4 1750
GenBank mRNA AF196344.1 - 5 1659

View Full Product Details