Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_023998.1 | NP_076488.1 | 1 | 638 | |
GenBank mRNA | AF227145.1 | - | 1 | 638 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | XM_006237482.1 | XP_006237544.1 | 1 | 698 |