Gene Transcripts

Gene Symbol
HHLA2
Entrez Gene ID
11148
Gene Name
HERV-H LTR-associating 2
Gene Aliases
B7-H5, B7-H7, B7H7, B7y
Chromosome Location
Chr. 3: 108296490 - 108378285 on Build GRCh38
UniGene ID
Hs.225968
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282556.1 NP_001269485.1 5 838
NM_001282557.1 NP_001269486.1 5 769
NM_001282558.1 NP_001269487.1 6 916
NM_001282559.1 NP_001269488.1 6 654
NM_007072.3 NP_009003.1 5 1029
XM_005247080.3 XP_005247137.1 6 899
XM_011512362.1 XP_011510664.1 6 899
XM_011512363.1 XP_011510665.1 5 829
XM_011512364.1 XP_011510666.1 5 780
XM_011512367.2 XP_011510669.1 6 899
XM_017005642.1 XP_016861131.1 5 829
GenBank mRNA AF126162.1 - 5 769
AK000692.1 - 4 764
AK027132.1 - 2 322
AK296644.1 - 6 597
AK315109.1 - 6 861
BC035971.1 - 5 1029 4606764

View Full Product Details