Gene Transcripts

Gene Symbol
Ankh
Entrez Gene ID
114506
Gene Name
ANKH inorganic pyrophosphate transport regulator
Gene Aliases
Ank
Chromosome Location
Chr. 2: 80131563 - 80256948 on Build Rnor_6.0
UniGene ID
Rn.211841
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053714.1 NP_446166.1 6 1004
GenBank mRNA AF393241.1 - 6 1004

View Full Product Details