Gene Transcripts

Gene Symbol
Filip1
Entrez Gene ID
246776
Gene Name
filamin A interacting protein 1
Gene Aliases
Filip
Chromosome Location
Chr. 8: 87268176 - 87419370 on Build Rnor_6.0
UniGene ID
Rn.44931
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145682.1 NP_663715.1 2 253
GenBank mRNA AB055759.1 - 2 253

View Full Product Details