Gene Transcripts

Gene Symbol
Prkaa2
Entrez Gene ID
78975
Gene Name
protein kinase, AMP-activated, alpha 2 catalytic subunit
Gene Aliases
Ampk, Ampka2
Chromosome Location
Chr. 5: 124568845 - 124642569 on Build Rnor_6.0
UniGene ID
Rn.64583
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023991.1 NP_076481.1 8 1406
XM_008763901.1 XP_008762123.1 7 3135
GenBank mRNA U12149.1 - 8 1591
Z29486.1 - 8 1406

View Full Product Details