Gene Transcripts
- Gene Symbol
- Taar6
- Entrez Gene ID
- 294124
- Gene Name
- trace amine-associated receptor 6
- Gene Aliases
- TRAR4, Ta4
- Chromosome Location
- Chr. 1: 22536199 - 22537236 on Build Rnor_6.0
- UniGene ID
- Rn.138139
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_175584.1 |
NP_783174.1
|
1 |
1014 |
|
View Full Product Details