Gene Transcripts

Gene Symbol
Mettl20
Entrez Gene ID
316976
Gene Name
methyltransferase like 20
Gene Aliases
Chromosome Location
Chr. 4: 183655972 - 183665757 on Build Rnor_6.0
UniGene ID
Rn.33980
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198772.1 NP_942067.1 3 467
XM_006237701.2 XP_006237763.1 2 401
GenBank mRNA BC061574.1 - 3 467

View Full Product Details