Gene Transcripts
- Gene Symbol
- Gucy2c
- Entrez Gene ID
- 25711
- Gene Name
- guanylate cyclase 2C
- Gene Aliases
- GCC
- Chromosome Location
- Chr. 4: 170659993 - 170740274 on Build Rnor_6.0
- UniGene ID
- Rn.10931
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_013170.1 |
NP_037302.1
|
1 |
151 |
|
GenBank mRNA |
M55636.1 |
- |
1 |
151 |
|
View Full Product Details