Gene Transcripts
- Gene Symbol
- Grem1
- Entrez Gene ID
- 50566
- Gene Name
- gremlin 1
- Gene Aliases
- Cktsf1b1, drm
- Chromosome Location
- Chr. 3: 105203309 - 105214989 on Build Rnor_6.0
- UniGene ID
- Rn.42929
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_019282.3 |
NP_062155.1
|
2 |
402 |
|
GenBank mRNA |
Y10019.1 |
- |
2 |
401 |
|
View Full Product Details