Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_012497.1 | NP_036629.1 | 3 | 317 | |
GenBank mRNA | BC059154.1 | - | 3 | 292 | |
BC099749.1 | - | 3 | 303 | 5599032 | |
FQ213506.1 | - | 3 | 320 | ||
FQ214420.1 | - | 3 | 343 | ||
FQ228893.1 | - | 3 | 317 | ||
X06984.1 | - | 3 | 317 |