Gene Transcripts

Gene Symbol
GTPBP4
Entrez Gene ID
23560
Gene Name
GTP binding protein 4
Gene Aliases
CRFG, NGB, NOG1
Chromosome Location
Chr. 10: 988409 - 1017768 on Build GRCh38
UniGene ID
Hs.215766
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012341.2 NP_036473.2 4 507
GenBank mRNA AF120334.1 - 4 459
AF325353.1 - 4 436
AK001548.1 - 4 454
AK001552.1 - 4 509
AK097093.1 - 3 350
AK222861.1 - 4 454
AK295231.1 - 3 350
AK301721.1 - 4 622
AK302219.1 - 3 312
BC033784.1 - 4 430
BC038975.2 - 4 468 5534986

View Full Product Details