Gene Transcripts

Gene Symbol
Efnb1
Entrez Gene ID
25186
Gene Name
ephrin B1
Gene Aliases
LERK2
Chromosome Location
Chr. X: 68891227 - 68904034 on Build Rnor_6.0
UniGene ID
Rn.44398
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017089.2 NP_058785.2 3 1225
GenBank mRNA BC061744.1 - 3 1225
U07560.1 - 3 1251

View Full Product Details