Gene Transcripts

Gene Symbol
Nsg1
Entrez Gene ID
25247
Gene Name
neuron specific gene family member 1
Gene Aliases
Bsmrb, Neep21, PEP1
Chromosome Location
Chr. 14: 77380264 - 77402007 on Build Rnor_6.0
UniGene ID
Rn.2865
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024128.3 NP_077042.2 5 812
GenBank mRNA BC060538.1 - 5 815

View Full Product Details