Gene Transcripts

Gene Symbol
Pld1
Entrez Gene ID
25096
Gene Name
phospholipase D1
Gene Aliases
Pld1a, Pld1b, Plda, Pldb
Chromosome Location
Chr. 2: 113651964 - 113849403 on Build Rnor_6.0
UniGene ID
Rn.11130
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030992.1 NP_112254.1 9 1066
XM_006232205.2 XP_006232267.1 9 1841
XM_006232206.2 XP_006232268.1 10 1494
XM_008760891.1 XP_008759113.1 10 1494
GenBank mRNA AB003170.1 - 9 958
AB003171.1 - 9 958
AF017251.1 - 8 910
U69550.1 - 9 1066
U88986.1 - 9 978

View Full Product Details