Gene Transcripts

Gene Symbol
Map1a
Entrez Gene ID
25152
Gene Name
microtubule-associated protein 1A
Gene Aliases
Mtap1a
Chromosome Location
Chr. 3: 113251864 - 113272193 on Build Rnor_6.0
UniGene ID
Rn.11402
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030995.1 NP_112257.1 4 573
XM_006234826.2 XP_006234888.1 5 937
XM_006234827.2 XP_006234889.1 5 937
XM_006234828.2 XP_006234890.1 4 841
XM_008762145.1 XP_008760367.1 4 577
GenBank mRNA M83196.1 - 4 573

View Full Product Details