Gene Transcripts

Gene Symbol
Musk
Entrez Gene ID
81725
Gene Name
muscle, skeletal, receptor tyrosine kinase
Gene Aliases
Nsk1
Chromosome Location
Chr. 5: 75392790 - 75498694 on Build Rnor_6.0
UniGene ID
Rn.10210
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031061.1 NP_112323.1 14 1911
GenBank mRNA U34985.1 - 14 1911

View Full Product Details