Gene Transcripts
- Gene Symbol
- AK5
- Entrez Gene ID
- 26289
- Gene Name
- adenylate kinase 5
- Gene Aliases
- AK6
- Chromosome Location
- Chr. 1: 77281908 - 77559969 on Build GRCh38
- UniGene ID
- Hs.559718
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_012093.3 |
NP_036225.2
|
8, 9 |
2015 |
|
NM_174858.2 |
NP_777283.1
|
8, 9 |
1404 |
|
XM_005270739.4 |
XP_005270796.1
|
8, 9 |
1251 |
|
XM_006710572.3 |
XP_006710635.1
|
8, 9 |
1182 |
|
XM_017001008.1 |
XP_016856497.1
|
8, 9 |
1199 |
|
XM_017001009.1 |
XP_016856498.1
|
8, 9 |
1907 |
|
XM_017001010.1 |
XP_016856499.1
|
8, 9 |
2234 |
|
XM_017001011.1 |
XP_016856500.1
|
8, 9 |
1017 |
|
XM_017001012.1 |
XP_016856501.1
|
8, 9 |
1473 |
|
GenBank mRNA |
AF062595.1 |
- |
1 |
22 |
|
AF445193.1 |
- |
8, 9 |
1404 |
|
AY171600.2 |
- |
8, 9 |
2020 |
|
BC012467.2 |
- |
3, 4 |
367 |
|
BC033896.1 |
- |
8, 9 |
2020 |
4818069 |
BC036666.1 |
- |
8, 9 |
1330 |
4816351 |
View Full Product Details