Gene Transcripts

Gene Symbol
OR2AG1
Entrez Gene ID
144125
Gene Name
olfactory receptor family 2 subfamily AG member 1 (gene/pseudogene)
Gene Aliases
OR11-79, OR2AG3
Chromosome Location
Chr. 11: 6785017 - 6786014 on Build GRCh38
UniGene ID
Hs.553608
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004489.2 NP_001004489.1 1 545
GenBank mRNA BC151143.1 - 1 545

View Full Product Details