Gene Transcripts

Gene Symbol
PADI1
Entrez Gene ID
29943
Gene Name
peptidyl arginine deiminase 1
Gene Aliases
HPAD10, PAD1, PDI, PDI1
Chromosome Location
Chr. 1: 17205123 - 17246006 on Build GRCh38
UniGene ID
Hs.412941
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013358.2 NP_037490.2 1 172
XM_011541307.1 XP_011539609.1 1 175
XM_017001103.1 XP_016856592.1 1 175
GenBank mRNA AB033768.2 - 1 163
AK026652.1 - 1 174
AK225541.1 - 1 174
AK314644.1 - 1 173
BC130574.1 - 1 151 40146605
BC136402.1 - 1 151 9020389

View Full Product Details