Gene Transcripts
- Gene Symbol
- SSX2IP
- Entrez Gene ID
- 117178
- Gene Name
- SSX family member 2 interacting protein
- Gene Aliases
- ADIP
- Chromosome Location
- Chr. 1: 84643707 - 84690757 on Build GRCh38
- UniGene ID
- Hs.22587
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001166293.1 |
NP_001159765.1
|
3, 4 |
497 |
|
NM_001166294.1 |
NP_001159766.1
|
2, 3 |
365 |
|
NM_001166295.1 |
NP_001159767.1
|
2, 3 |
493 |
|
NM_001166417.1 |
NP_001159889.1
|
4, 5 |
675 |
|
NM_014021.3 |
NP_054740.3
|
4, 5 |
547 |
|
XM_005270427.1 |
XP_005270484.1
|
3, 4 |
436 |
|
XM_005270428.1 |
XP_005270485.1
|
3, 4 |
394 |
|
XM_005270429.1 |
XP_005270486.1
|
3, 4 |
436 |
|
XM_006710331.1 |
XP_006710394.1
|
3, 4 |
354 |
|
XM_011540610.1 |
XP_011538912.1
|
4, 5 |
444 |
|
XM_011540612.2 |
XP_011538914.1
|
4, 5 |
485 |
|
XM_017000234.1 |
XP_016855723.1
|
3, 4 |
425 |
|
XM_017000235.1 |
XP_016855724.1
|
3, 4 |
403 |
|
XM_017000236.1 |
XP_016855725.1
|
3, 4 |
419 |
|
XM_017000237.1 |
XP_016855726.1
|
2, 3 |
208 |
|
GenBank mRNA |
AB023140.1 |
- |
3, 4 |
485 |
|
AK021515.1 |
- |
2 |
1069 |
|
AK093173.1 |
- |
4, 5 |
432 |
|
AK292475.1 |
- |
3, 4 |
385 |
|
AK294057.1 |
- |
2, 3 |
252 |
|
AK316472.1 |
- |
2, 3 |
271 |
|
AL133046.1 |
- |
4, 5 |
487 |
|
AY367055.1 |
- |
4, 5 |
410 |
|
AY651262.1 |
- |
1, 2 |
70 |
|
BC033637.1 |
- |
3, 4 |
372 |
5578582 |
BC035580.1 |
- |
2, 3 |
306 |
|
BC064389.1 |
- |
3, 4 |
461 |
5745142 |
View Full Product Details