Gene Transcripts

Gene Symbol
OSGIN1
Entrez Gene ID
29948
Gene Name
oxidative stress induced growth inhibitor 1
Gene Aliases
BDGI, OKL38
Chromosome Location
Chr. 16: 83953222 - 83966332 on Build GRCh38
UniGene ID
Hs.128055
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_182981.2 NP_892026.1 6 1920
GenBank mRNA AF191740.1 - 4 1578
AY258066.1 - 7 2371
AY258067.1 - 6 2211
AY258068.1 - 6 1901

View Full Product Details