Gene Transcripts

Gene Symbol
SLC27A6
Entrez Gene ID
28965
Gene Name
solute carrier family 27 member 6
Gene Aliases
ACSVL2, FACVL2, FATP6, VLCS-H1
Chromosome Location
Chr. 5: 128965127 - 129033642 on Build GRCh38
UniGene ID
Hs.49765
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001017372.2 NP_001017372.1 10 2836
NM_001317984.1 NP_001304913.1 11 2660
NM_014031.4 NP_054750.1 11 2611
GenBank mRNA AF064254.1 - 10 1978
AK313456.1 - 11 2259
AL833117.1 - 10 2408
BC041945.1 - 10 2835 5301306
BX537383.1 - 11 2221
CR457129.1 - 10 1825

View Full Product Details