Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_014077.3 | NP_054796.1 | 2, 3 | 257 | |
GenBank mRNA | AF151902.1 | - | 2, 3 | 216 | |
AK312058.1 | - | 2, 3 | 240 | ||
AL050157.1 | - | 2, 3 | 235 | ||
BC000639.1 | - | 2, 3 | 235 | 3345441 | |
BC004447.1 | - | 1, 2 | 87 | ||
BC017286.1 | - | 2, 3 | 218 | 4892079 | |
BG749750.1 | - | 1, 2 | 384 |