Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001243225.1 | NP_001230154.1 | 3 | 274 | |
NM_001282327.1 | NP_001269256.1 | 5 | 672 | ||
NM_001282328.1 | NP_001269257.1 | 5 | 719 | ||
NM_014303.3 | NP_055118.1 | 3 | 274 | ||
XM_006724173.1 | XP_006724236.1 | 3 | 230 | ||
GenBank mRNA | AB451380.1 | - | 3 | 167 | |
AK096320.1 | - | 5 | 691 | ||
AK127185.1 | - | 5 | 673 | ||
AK315518.1 | - | 3 | 241 | ||
AU098520.1 | - | 3 | 274 | ||
AU130886.1 | - | 3 | 241 | ||
BC032489.1 | - | 3 | 231 | 5416478 | |
CR456539.1 | - | 3 | 192 | ||
U78310.1 | - | 3 | 225 |