Gene Transcripts

Gene Symbol
FTCD
Entrez Gene ID
10841
Gene Name
formimidoyltransferase cyclodeaminase
Gene Aliases
LCHC1
Chromosome Location
Chr. 21: 46135981 - 46156481 on Build GRCh38
UniGene ID
Hs.415846
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001320412.1 NP_001307341.1 6 759
NM_006657.2 NP_006648.1 6 759
NM_206965.1 NP_996848.1 6 759
XM_006723961.3 XP_006724024.2 6 1050
XM_006723962.3 XP_006724025.2 6 1658
XM_011529434.2 XP_011527736.1 6 1673
XM_011529435.2 XP_011527737.1 6 1057
XM_011529436.2 XP_011527738.1 6 1658
XM_011529437.2 XP_011527739.1 6 1052
XM_011529439.2 XP_011527741.1 4 462
XM_011529440.2 XP_011527742.1 6 1044
GenBank mRNA AF169017.1 - 6 759
AF289021.1 - 6 759
AF289022.1 - 6 759
AF289023.1 - 6 759
AF289024.1 - 6 784
BC052248.1 - 6 755 6270063
BC136383.1 - 6 751 9020370
BC136395.1 - 6 751 9020382

View Full Product Details