Gene Transcripts

Gene Symbol
PROSC
Entrez Gene ID
11212
Gene Name
proline synthetase cotranscribed homolog (bacterial)
Gene Aliases
Chromosome Location
Chr. 8: 37761753 - 37779768 on Build GRCh38
UniGene ID
Hs.304792
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007198.3 NP_009129.1 3, 4 314
XM_017013001.1 XP_016868490.1 3, 4 1144
GenBank mRNA AK021923.1 - 3, 4 182
AK291135.1 - 3, 4 252
AL136616.1 - 3, 4 294
BC012334.1 - 3, 4 304 3546307
CR533532.1 - 3, 4 237
DA086552.1 - 3, 4 314

View Full Product Details