Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001267812.1 | NP_001254741.1 | 6 | 955 | |
NM_001267814.1 | NP_001254743.1 | 4 | 692 | ||
NM_020246.3 | NP_064631.2 | 6 | 955 | ||
XM_005250502.2 | XP_005250559.1 | 4 | 691 | ||
XM_005250504.4 | XP_005250561.1 | 1 | 38 | ||
XM_006716054.2 | XP_006716117.1 | 4 | 673 | ||
XM_006716055.2 | XP_006716118.1 | 4 | 528 | ||
XM_011516414.1 | XP_011514716.1 | 3 | 361 | ||
GenBank mRNA | AB033284.1 | - | 6 | 882 | |
AF284422.1 | - | 6 | 922 | ||
AK024420.1 | - | 3 | 1215 | ||
AK024421.1 | - | 2 | 1772 | ||
AK024466.1 | - | 1 | 137 | ||
AK024494.1 | - | 1 | 2080 | ||
AK090458.1 | - | 2 | 1081 | ||
AK301411.1 | - | 6 | 955 | ||
BC000154.2 | - | 4 | 674 | 2900382 |