Gene Transcripts

Gene Symbol
RHEBL1
Entrez Gene ID
121268
Gene Name
Ras homolog enriched in brain like 1
Gene Aliases
RHEBL1c
Chromosome Location
Chr. 12: 49064676 - 49070682 on Build GRCh38
UniGene ID
Hs.159013
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001303126.1 NP_001290055.1 8 1066
NM_144593.2 NP_653194.1 8 998
NR_130123.1 7 930
GenBank mRNA AK098663.1 - 8 997
AY327412.1 - 8 998
AY509932.1 - 8 1064
BC014155.2 - 7 852
BC027482.1 - 8 965 4396827

View Full Product Details