Gene Transcripts

Gene Symbol
OR5A1
Entrez Gene ID
219982
Gene Name
olfactory receptor family 5 subfamily A member 1
Gene Aliases
OR11-249, OR5A1P, OST181
Chromosome Location
Chr. 11: 59443169 - 59444116 on Build GRCh38
UniGene ID
Hs.553649
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004728.1 NP_001004728.1 1 281
GenBank mRNA BC137060.1 - 1 306
BC137061.1 - 1 306 9021058

View Full Product Details