Gene Transcripts

Gene Symbol
ARMC8
Entrez Gene ID
25852
Gene Name
armadillo repeat containing 8
Gene Aliases
GID5, HSPC056, S863-2, VID28
Chromosome Location
Chr. 3: 138187248 - 138298389 on Build GRCh38
UniGene ID
Hs.744868
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001267042.2 NP_001253971.1 5, 6 735
NM_014154.4 NP_054873.2 6, 7 811
NM_015396.5 NP_056211.2 6, 7 811
NM_213654.3 NP_998819.1 6, 7 865
XM_006713564.3 XP_006713627.1 5, 6 730
XM_006713565.2 XP_006713628.1 6, 7 771
XM_006713566.3 XP_006713629.1 4, 5 625
XM_006713567.3 XP_006713630.1 5, 6 730
XM_011512632.1 XP_011510934.1 5, 6 678
XM_011512633.1 XP_011510935.1 5, 6 732
XM_017006095.1 XP_016861584.1 6, 7 733
XM_017006096.1 XP_016861585.1 4, 5 625
XM_017006097.1 XP_016861586.1 5, 6 679
XM_017006098.1 XP_016861587.1 4, 5 625
XM_017006099.1 XP_016861588.1 5, 6 716
XM_017006100.1 XP_016861589.1 5, 6 730
GenBank mRNA AF109184.1 - 6, 7 564
AF161541.1 - 6, 7 533
AK001721.1 - 4, 5 373
AK289577.1 - 6, 7 507
AK296757.1 - 5, 6 466
BC013424.1 - 6, 7 565 3892143
BC032661.1 - 6, 7 675 5530425
BC041699.1 - 6, 7 490 5240423
CX167429.1 - 5, 6 440

View Full Product Details