Gene Transcripts

Gene Symbol
OR8B4
Entrez Gene ID
283162
Gene Name
olfactory receptor family 8 subfamily B member 4 (gene/pseudogene)
Gene Aliases
OR11-315, OR8B4P
Chromosome Location
Chr. 11: 124423942 - 124424871 on Build GRCh38
UniGene ID
Hs.553671
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005196.1 NP_001005196.1 1 148
GenBank mRNA BC136865.1 - 1 170
BC136866.1 - 1 170 9020857

View Full Product Details